Toluene-α-sulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: Benzylsulphonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride

A578645-10G A578645-25G
77144-702EA 6.18 USD
77144-702 77569-102
Toluene-α-sulphonyl chloride 98%
Toluene-α-sulphonyl chloride

Precaución: For research use only.

Formula: C₇H₇ClO₂S
MDL Number: MFCD00007455
CAS Number: 1939-99-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR