3-Chloro-4-methylbenzenesulfonyl chloride 96%

Supplier: Ambeed
Danger

Synonyms: 3-Chlor-4-methylbenzenesulphonyl chloride, 2-Chloro-p-toluenesulphochloride, 3-Chloro-4-methylbenzene-1-sulphonyl chloride

A576047-1G A576047-25G A576047-5G
77144-686EA 10.61 USD
77144-686 77144-688 77144-690
3-Chloro-4-methylbenzenesulfonyl chloride 96%
3-Chloro-4-methylbenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
MDL Number: MFCD00203987
CAS Number: 42413-03-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR