(S)-(+)-1-Methoxy-2-propanol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-1-Methoxypropan-2-ol, (S)-(+)-Propylene glycol 1-monomethyl ether
A575359-1G
A575359-25G
A575359-5G
77144-670EA
8.49
USD
77144-670
77144-672
77144-674
(S)-(+)-1-Methoxy-2-propanol 98%
(S)-(+)-1-Methoxy-2-propanol
Precaución: for Research Use Only
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol |
MDL Number:
MFCD01632588 CAS Number: 26550-55-0 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...