1-Ethynylcyclohexanol 98%

Supplier: Ambeed
Danger

Synonyms: 1-Ethynyl-1-cyclohexanol

A574905-25G A574905-100G
77144-666EA 12.36 USD
77144-666 77686-464
1-Ethynylcyclohexanol 98%
1-Ethynylcyclohexanol

Precaución: for Research Use Only

Formula: C₈H₁₂O
MDL Number: MFCD00003858
CAS Number: 78-27-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR