2-Methyl-5-nitrobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Nitrotoluene-2-sulphonyl chloride, 2-Methyl-5-nitrobenzene-1-sulphonyl chloride

A572721-10G A572721-25G A572721-5G A572721-1G
77144-650EA 35.01 USD
77144-650 77144-652 77144-654 77747-185
2-Methyl-5-nitrobenzenesulfonyl chloride 98%
2-Methyl-5-nitrobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆ClNO₄S
MW: 235.65 g/mol
MDL Number: MFCD00051695
CAS Number: 121-02-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR