N,N-Di-n-butyllactamide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N-Dibutyl-2-hydroxypropanamide
A572139-100MG
A572139-1G
A572139-250MG
77144-638EA
14.42
USD
77144-638
77144-640
77144-642
N,N-Di-n-butyllactamide 95%
N,N-Di-n-butyllactamide
Precaución: for Research Use Only
Formula:
C₁₁H₂₃NO₂ MW: 201.3058 g/mol |
MDL Number:
MFCD00086703 CAS Number: 6288-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...