3,6-Dichloro-2-methoxypyridine 96%

Supplier: Ambeed
Warning

A570351-100MG A570351-1G A570351-250MG
77144-630EA 43.5 USD
77144-630 77144-632 77144-634
3,6-Dichloro-2-methoxypyridine 96%
3,6-Dichloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.016 g/mol
MDL Number: MFCD13185545
CAS Number: 1214391-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR