3-(Bromoacetyl)-2H-chromen-2-one 98%

Supplier: Ambeed
Danger

A568605-1G A568605-5G
77144-618EA 22.66 USD
77144-618 77144-620
3-(Bromoacetyl)-2H-chromen-2-one 98%
3-(Bromoacetyl)-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₁H₇BrO₃
MDL Number: MFCD00488331
CAS Number: 29310-88-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR