3,5-Diiodobenzaldehyde 98%

Supplier: Ambeed
Warning

A567383-100MG A567383-1G A567383-250MG
77144-604EA 16.47 USD
77144-604 77144-606 77144-608
3,5-Diiodobenzaldehyde 98%
3,5-Diiodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄I₂O
MDL Number: MFCD07779028
CAS Number: 17352-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR