2-(1H-Pyrazol-1-yl)-5-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A565554-100MG A565554-1G A565554-250MG
77144-592EA 67.9 USD
77144-592 77144-594 77144-596
2-(1H-Pyrazol-1-yl)-5-(trifluoromethyl)pyrimidine 97%
2-(1H-Pyrazol-1-yl)-5-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₈H₅F₃N₄
MW: 214.14 g/mol
MDL Number: MFCD22574210
CAS Number: 1398504-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR