(1S,2S)-Boc-Achc 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid

A562269-100MG A562269-250MG
77144-582EA 102.91 USD
77144-582 77617-386
(1S,2S)-Boc-Achc 98%
(1S,2S)-Boc-Achc

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
MDL Number: MFCD04974226
CAS Number: 488703-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR