(Methylthio)acetic acid 95%
Supplier: Ambeed
|
Warning
|
A561760-1G
A561760-250MG
A561760-25G
A561760-5G
77144-574EA
11.34
USD
77144-574
77144-576
77144-578
77144-580
(Methylthio)acetic acid 95%
(Methylthio)acetic acid
Precaución: for Research Use Only
Formula:
C₃H₆O₂S |
MDL Number:
MFCD00075444 CAS Number: 2444-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...