(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol 98%

Supplier: Ambeed
Warning

Synonyms: L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

A561194-10G A561194-25G
77144-570EA 6.18 USD
77144-570 77144-572
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol 98%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄
MDL Number: MFCD00007359
CAS Number: 2964-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR