4-Nitro-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A560445-100MG A560445-1G A560445-250MG
77144-560EA 81.69 USD
77144-560 77144-562 77144-564
4-Nitro-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
4-Nitro-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃O₂
MW: 165.15 g/mol
MDL Number: MFCD27991797
CAS Number: 1822922-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR