Methyl-m-toluate 98%

Supplier: Ambeed
Warning

Synonyms: m-Toluic acid methyl ester, Methyl 3-methylbenzoate, 3-Toluic acid methyl ester, Methyl-3-methylbenzoate, 3-Methylbenzoic acid methyl ester

A559087-100G A559087-25G A559087-500G
77144-552EA 46.68 USD
77144-552 77144-554 77144-556
Methyl-m-toluate 98%
Methyl-m-toluate

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00008436
CAS Number: 99-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR