3,5-Difluoro-3'-methoxy-[1,1'-biphenyl]-4-amine 97%
Supplier: Ambeed
|
Warning
|
A558502-1G
A558502-250MG
77144-546EA
150.65
USD
77144-546
77144-548
3,5-Difluoro-3'-methoxy-[1,1'-biphenyl]-4-amine 97%
3,5-Difluoro-3'-methoxy-[1,1'-biphenyl]-4-amine
Precaución: for Research Use Only
Formula:
C₁₃H₁₁F₂NO MW: 235.22 g/mol |
MDL Number:
MFCD14585527 CAS Number: 867288-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...