2,2,2-Trifluoro-1-(m-tolyl)ethanamine 97%

Supplier: Ambeed
Danger

A554596-1G A554596-250MG
77144-524EA 589.87 USD
77144-524 77144-526
2,2,2-Trifluoro-1-(m-tolyl)ethanamine 97%
2,2,2-Trifluoro-1-(m-tolyl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₀F₃N
MW: 189.17 g/mol
MDL Number: MFCD07374604
CAS Number: 886368-58-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR