2,4-Difluoro-5-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2,4-difluorobenzotrifluoride

A553608-100MG A553608-1G A553608-250MG
77144-510EA 16.98 USD
77144-510 77144-512 77144-514
2,4-Difluoro-5-(trifluoromethyl)aniline 97%
2,4-Difluoro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄F₅N
MW: 197.1054 g/mol
MDL Number: MFCD01631613
CAS Number: 261944-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR