2-(4-Bromophenyl)-2-oxoacetaldehyde 95%

Supplier: Ambeed
Warning

A552399-100MG A552399-250MG
77144-504EA 56.64 USD
77144-504 77144-506
2-(4-Bromophenyl)-2-oxoacetaldehyde 95%
2-(4-Bromophenyl)-2-oxoacetaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrO₂
MDL Number: MFCD00998793
CAS Number: 5195-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR