Glimepiride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Ethyl-4-methyl-N-(4-(N-((rel-(1r,4r)-4-methylcyclohexyl)carbamoyl)sulphamoyl)phenethyl)-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide

A552202-1MG A552202-250MG A552202-5G A552202-1G A552202-25G
77144-492EA 5.31 USD
77144-492 77144-494 77144-496 77617-356 77686-418
Glimepiride 97%
Glimepiride

Precaución: for Research Use Only

Formula: C₂₄H₃₄N₄O₅S
MW: 490.62 g/mol
MDL Number: MFCD00878417
CAS Number: 93479-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR