2-(Benzyloxy)-1-(2-fluorophenyl)ethanone 97%

Supplier: Ambeed
Warning

A551021-1G A551021-250MG
77144-488EA 185.66 USD
77144-488 77144-490
2-(Benzyloxy)-1-(2-fluorophenyl)ethanone 97%
2-(Benzyloxy)-1-(2-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
MW: 244.26 g/mol
MDL Number: MFCD15519617
CAS Number: 1402742-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR