tert-Butyl 3-(aminomethyl)-4-methylpiperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A549719-100MG A549719-250MG
77144-476EA 87.55 USD
77144-476 77144-478
tert-Butyl 3-(aminomethyl)-4-methylpiperazine-1-carboxylate 97%
tert-Butyl 3-(aminomethyl)-4-methylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₃N₃O₂
MW: 229.31 g/mol
MDL Number: MFCD22035431
CAS Number: 1369237-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR