(5-Bromo-2,3-difluorophenyl)boronic acid 97%

Supplier: Ambeed
Warning

A549617-1G A549617-250MG A549617-5G
77144-470EA 82.4 USD
77144-470 77144-472 77144-474
(5-Bromo-2,3-difluorophenyl)boronic acid 97%
(5-Bromo-2,3-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrF₂O₂
MDL Number: MFCD19237164
CAS Number: 2096339-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR