2,7-Di(9H-carbazol-9-yl)-9,9'-spirobi[fluorene] 98%

Supplier: Ambeed
Warning

A547377-1G A547377-250MG
77144-456EA 55.61 USD
77144-456 77144-458
2,7-Di(9H-carbazol-9-yl)-9,9'-spirobi[fluorene] 98%
2,7-Di(9H-carbazol-9-yl)-9,9'-spirobi[fluorene]

Precaución: for Research Use Only

Formula: C₄₉H₃₀N₂
MDL Number: MFCD12022464
CAS Number: 924899-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR