(S)-N-Glycidylphthalimide 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione, (S)-N-(2,3-Epoxypropyl)phthalimide

A546157-1000G A546157-100G A546157-10G A546157-25G A546157-500G A546157-5G
77144-444EA 324.45 USD
77144-444 77144-446 77144-448 77144-450 77144-452 77144-454
(S)-N-Glycidylphthalimide 97%
(S)-N-Glycidylphthalimide

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MDL Number: MFCD04973350
CAS Number: 161596-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR