(R)-2-((tert-Butoxycarbonyl)amino)-2-phenylpropanoic acid 95%

Supplier: Ambeed
Warning

A544297-100MG A544297-250MG
77144-438EA 96.55 USD
77144-438 77144-440
(R)-2-((tert-Butoxycarbonyl)amino)-2-phenylpropanoic acid 95%
(R)-2-((tert-Butoxycarbonyl)amino)-2-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.30 g/mol
MDL Number: MFCD14585172
CAS Number: 365442-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR