4-Bromo-2-chloro-6-(trifluoromethyl)pyridin-3-amine 97%

Supplier: Ambeed
Danger

A543431-100MG A543431-1G
77144-428EA 28.84 USD
77144-428 77144-430
4-Bromo-2-chloro-6-(trifluoromethyl)pyridin-3-amine 97%
4-Bromo-2-chloro-6-(trifluoromethyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₃BrClF₃N₂
MW: 275.45 g/mol
MDL Number: MFCD22199859
CAS Number: 947146-74-9
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR