2-(2-Bromo-6-chlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A543417-100MG A543417-1G A543417-250MG
77144-422EA 21.63 USD
77144-422 77144-424 77144-426
2-(2-Bromo-6-chlorophenyl)acetic acid 97%
2-(2-Bromo-6-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD18390580
CAS Number: 37777-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR