4-Bromo-2-fluoro-6-nitroanisole 98%

Supplier: Ambeed
Warning

A542316-1G A542316-5G
77144-412EA 22.66 USD
77144-412 77144-414
4-Bromo-2-fluoro-6-nitroanisole 98%
4-Bromo-2-fluoro-6-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₃
MDL Number: MFCD00013374
CAS Number: 74266-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR