(R)-Ethyl-2-hydroxypropanoate 98%

Supplier: Ambeed
Danger

Synonyms: (+)-Ethyl d-lactate, (R)-Ethyl 2-hydroxypropanoate

A540265-10G A540265-1G A540265-25G A540265-5G A540265-100G
77144-390EA 8.24 USD
77144-390 77144-392 77144-394 77144-396 77686-389
(R)-Ethyl-2-hydroxypropanoate 98%
(R)-Ethyl-2-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MDL Number: MFCD00077825
CAS Number: 7699-00-5
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR