(3S,4S)-3-Amino-1-chloro-4-methylhexan-2-one hydrochloride 96%

Supplier: Ambeed
Warning

A537717-100MG A537717-1G A537717-250MG A537717-50MG
77144-368EA 23.34 USD
77144-368 77144-370 77144-372 77144-374
(3S,4S)-3-Amino-1-chloro-4-methylhexan-2-one hydrochloride 96%
(3S,4S)-3-Amino-1-chloro-4-methylhexan-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅Cl₂NO
MW: 200.10 g/mol
MDL Number: MFCD19443520
CAS Number: 59079-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR