(5-Chloro-2,3-difluorophenyl)boronic acid 97%
Supplier: Ambeed
|
Warning
|
A537318-100MG
A537318-1G
A537318-250MG
77144-362EA
38.12
USD
77144-362
77144-364
77144-366
(5-Chloro-2,3-difluorophenyl)boronic acid 97%
(5-Chloro-2,3-difluorophenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₆H₄BClF₂O₂ |
MDL Number:
MFCD28751294 CAS Number: 2377610-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...