3-Bromo-6-fluorobenzene -1,2-diamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,3-diamino-4-fluorobenzene

A536996-100MG A536996-250MG
77144-358EA 105.03 USD
77144-358 77144-360
3-Bromo-6-fluorobenzene -1,2-diamine 95%
3-Bromo-6-fluorobenzene -1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₆BrFN₂
MW: 205.028 g/mol
MDL Number: MFCD28154969
CAS Number: 1805502-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR