tert-Butyl 2-propylpiperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A535915-100MG A535915-1G A535915-250MG
77144-342EA 44.56 USD
77144-342 77144-344 77144-346
tert-Butyl 2-propylpiperazine-1-carboxylate 98%
tert-Butyl 2-propylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MW: 228.33 g/mol
MDL Number: MFCD08273955
CAS Number: 1027511-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR