2-Chloro-4-fluoroanisole 98%

Supplier: Ambeed
Warning

A535528-100G A535528-1G A535528-5G
77144-336EA 318.27 USD
77144-336 77144-338 77144-340
2-Chloro-4-fluoroanisole 98%
2-Chloro-4-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD00672965
CAS Number: 2267-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR