(3-Chloro-5-(2-methoxyethoxy)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A534369-100MG A534369-1G A534369-250MG
77144-328EA 40.18 USD
77144-328 77144-330 77144-332
(3-Chloro-5-(2-methoxyethoxy)phenyl)boronic acid 97%
(3-Chloro-5-(2-methoxyethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₄
MW: 230.45 g/mol
MDL Number: MFCD21609544
CAS Number: 2096335-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR