(R)-(+)-3-Methoxy-α-methylbenzylamine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-1-(3-Methoxyphenyl)ethanamine, (R)-(+)-1-(3-Methoxyphenyl)ethylamine
A534226-10G
A534226-1G
A534226-250MG
A534226-5G
77144-322EA
369.2
USD
77144-322
77144-324
77144-326
77747-090
(R)-(+)-3-Methoxy-α-methylbenzylamine 98%
(R)-(+)-3-Methoxy-α-methylbenzylamine
Precaución: for Research Use Only
Formula:
C₉H₁₃NO MW: 151.21 g/mol |
MDL Number:
MFCD00671655 CAS Number: 88196-70-7 UN: 2922 ADR: 8,6.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...