(R)-(+)-1-(2-Naphthyl)ethylamine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(2-Naphthyl)ethylamine

A534082-10G A534082-1G A534082-25G A534082-5G
77144-314EA 254.62 USD
77144-314 77144-316 77144-318 77144-320
(R)-(+)-1-(2-Naphthyl)ethylamine 98%
(R)-(+)-1-(2-Naphthyl)ethylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
MDL Number: MFCD00085367
CAS Number: 3906-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR