rel-(1r,4r)-4-((1,1-Dimethylethyl)sulfonamido)-N-(5-(trifluoromethyl)pyridin-2-yl)cyclohexane-1-carboxamide 98%

Supplier: Ambeed
Warning

A533511-10MG A533511-1MG A533511-25MG A533511-50MG A533511-5MG
77144-304EA 113.31 USD
77144-304 77144-306 77144-308 77144-310 77144-312
rel-(1r,4r)-4-((1,1-Dimethylethyl)sulfonamido)-N-(5-(trifluoromethyl)pyridin-2-yl)cyclohexane-1-carboxamide 98%
rel-(1r,4r)-4-((1,1-Dimethylethyl)sulfonamido)-N-(5-(trifluoromethyl)pyridin-2-yl)cyclohexane-1-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₂₄F₃N₃O₃S
MDL Number: MFCD11973642
CAS Number: 342577-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR