(S)-1-(2-Methoxyphenyl)ethylamine 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-1-(2-Methoxyphenyl)ethanamine
A532682-100MG
A532682-1G
A532682-5G
A532682-250MG
77144-290EA
15.45
USD
77144-290
77144-292
77144-294
77617-268
(S)-1-(2-Methoxyphenyl)ethylamine 95%
(S)-1-(2-Methoxyphenyl)ethylamine
Precaución: for Research Use Only
Formula:
C₉H₁₃NO |
MDL Number:
MFCD00671658 CAS Number: 68285-24-5 UN: 2922 ADR: 8,6.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...