2,3-Dimethoxyterephthalaldehyde 95%

Supplier: Ambeed
Warning

A532477-100MG A532477-250MG A532477-1G
77144-286EA 79.57 USD
77144-286 77144-288 77551-612
2,3-Dimethoxyterephthalaldehyde 95%
2,3-Dimethoxyterephthalaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD18072900
CAS Number: 179693-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR