4-Methoxyphenyl-4-n-propylcyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxyphenyl 4-propylcyclohexane-1-carboxylate, 4-Methoxyphenyl 4-propylcyclohexanecarboxylate

A530979-100G A530979-25G A530979-5G
77144-272EA 63.86 USD
77144-272 77144-274 77144-276
4-Methoxyphenyl-4-n-propylcyclohexanecarboxylate 97%
4-Methoxyphenyl-4-n-propylcyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₄O₃
MDL Number: MFCD11053431
CAS Number: 67589-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR