1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane 98%

Supplier: Ambeed
Warning

Synonyms: (3-Glycidoxypropyl)bis(trimethylsiloxy)methylsilane

A530453-1G A530453-25G A530453-5G
77144-262EA 8.49 USD
77144-262 77144-264 77144-266
1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane 98%
1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane

Precaución: for Research Use Only

Formula: C₁₃H₃₂O₄Si₃
MW: 336.65 g/mol
MDL Number: MFCD00053926
CAS Number: 7422-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR