3,5-Diiodosalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Diiodo-2-hydroxybenzaldehyde

A530403-1G A530403-5G A530403-25G A530403-100G
77144-258EA 10.3 USD
77144-258 77144-260 77569-018 77686-370
3,5-Diiodosalicylaldehyde 98%
3,5-Diiodosalicylaldehyde

Precaución: For research use only.

Formula: C₇H₄I₂O₂
MDL Number: MFCD00003321
CAS Number: 2631-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR