(2-Hydroxy-5-((trifluoromethyl)thio)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A528911-100MG A528911-250MG
77144-252EA 40.18 USD
77144-252 77144-254
(2-Hydroxy-5-((trifluoromethyl)thio)phenyl)boronic acid 97%
(2-Hydroxy-5-((trifluoromethyl)thio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₃S
MDL Number: MFCD19684110
CAS Number: 2377609-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR