(R)-4-Benzyloxazolidine-2-thione 97%

Supplier: Ambeed
Warning

A528677-100MG A528677-1G A528677-250MG
77144-246EA 12.36 USD
77144-246 77144-248 77144-250
(R)-4-Benzyloxazolidine-2-thione 97%
(R)-4-Benzyloxazolidine-2-thione

Precaución: for Research Use Only

Formula: C₁₀H₁₁NOS
MDL Number: MFCD06658222
CAS Number: 190970-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR