(S)-(-)-2-Acetoxypropionyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: (S)-(-)-2-acetoxypropionylchloride, (S)-(-)-O-Acetyllactoyl Chloride

A528435-1G A528435-25G A528435-5G
77144-240EA 41.38 USD
77144-240 77144-242 77144-244
(S)-(-)-2-Acetoxypropionyl chloride 98%
(S)-(-)-2-Acetoxypropionyl chloride

Precaución: for Research Use Only

Formula: C₅H₇ClO₃
MW: 150.56 g/mol
MDL Number: MFCD00145252
CAS Number: 36394-75-9
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR