1,2-Bis(carboxymethylthio)ethane 98%

Supplier: Ambeed
Warning

Synonyms: (Ethylenedithio)diacetic acid, (Ethylenedithio)diaceticacid

A528147-100G A528147-25G A528147-5G
77144-234EA 181.42 USD
77144-234 77144-236 77144-238
1,2-Bis(carboxymethylthio)ethane 98%
1,2-Bis(carboxymethylthio)ethane

Precaución: for Research Use Only

Formula: C₆H₁₀O₄S₂
MW: 210.26 g/mol
MDL Number: MFCD00021771
CAS Number: 7244-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR