5-Hexyl-2,2':5',2''-terthiophene 97%

Supplier: Ambeed
Warning

A526253-100MG A526253-250MG
77144-218EA 44.28 USD
77144-218 77144-220
5-Hexyl-2,2':5',2''-terthiophene 97%
5-Hexyl-2,2':5',2''-terthiophene

Precaución: for Research Use Only

Formula: C₁₈H₂₀S₃
MDL Number: MFCD28291932
CAS Number: 151271-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR