3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A525095-1G A525095-5G
77144-208EA 13.8 USD
77144-208 77144-210
3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine 98%
3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MDL Number: MFCD00153096
CAS Number: 175136-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR